VZV gE
Supplier: Biorbyt
Certificates
About this item
Specifications
- Lagerbedingungen:Five years frozen. One month in solution at room temperature.
- Protein/Peptid-Name:VZV gE
- Purity:Varicella protein is >95% pure as determined by 10% PAGE (coomassie staining). Varicella was purified by proprietary chromatographic technique.
- Zusammensetzung:25mM Tris-Hcl pH 8.0, 1mM EDTA and 50% glycerol.
- Validierte Anwendungen:SDS-PAGE