ContactUsLinkComponent
Toxoplasma MIC3
Toxoplasma MIC3
Supplier:  Biorbyt
Certificates
Toxoplasma MIC3
Supplier:  Biorbyt
Specifications
  • Lagerbedingungen:
    Five years frozen. One month in solution at room temperature.
  • Protein/Peptid-Name:
    Toxoplasma MIC3
  • Purity:
    Toxoplasma protein is >95% pure as determined by 10% PAGE (coomassie staining). Toxoplasama protein was purified by proprietary chromatographic technique.
  • Zusammensetzung:
    50mM Tris-HCl, 1.5 urea and 50% glycerol.
  • Validierte Anwendungen:
    SDS-PAGE
Specifications
Related Information
Frequently Bought Together