ContactUsLinkComponent
SARS S(M)
SARS S(M)
Supplier:  Biorbyt
Certificates
SARS S(M)
Supplier:  Biorbyt
Specifications
  • Lagerbedingungen:
    Five years frozen. One month at 4 °C.
  • Protein/Peptid-Name:
    SARS S(M)
  • Purity:
    SARS Mosaic protein is >95% pure as determined by 10% PAGE (Coomassie staining). SARS Mosaic protein was purified by proprietary chromatographic technique.
  • Zusammensetzung:
    25mM Tris-HCl, 0.4% sarcosyl, 0,25% Triton C100 and 50% glycerol.
  • Validierte Anwendungen:
    SDS-PAGE
Specifications
Related Information
Frequently Bought Together