HCV NS3 genotype-5a
Supplier: Biorbyt
Certificates
About this item
Specifications
- Lagerbedingungen:Five years frozen. One month in solution at room temperature.
- Protein/Peptid-Name:HCV NS3 Genotype-5a
- Purity:HCV NS3 Genotype-5a protein is >95% pure as determined by 10% PAGE (coomassie staining). HCV NS3 Genotype-5a protein was purified by proprietary chromatographic technique.
- Zusammensetzung:25mM Tris-HCl, 1mM EDTA, 1.5M urea 50% glycerol.
- Validierte Anwendungen:SDS-PAGE