Human fetuin A (from plasma)
: Biorbyt
- Ursprung:Plasma
- Spezies:Human
- Lagerbedingungen:Store the lyophilized protein at −20 °C. Aliquot the product after reconstitution to avoid repeated freezing/thawing cycles. Reconstituted protein can be stored at 4 °C for limited period of time; it does not show any change after two weeks at 4 °C. The lyophilized protein remains stable until the expiry date when stored at −20 °C.
- Proteinsynonym:Alpha-2-HS-glycoprotein
- Protein/Peptid-Name:Fetuin A
- Purity:Greater than 95% as determined by SDS-PAGE.
- Solubility:It is recommended to add deionized water to prepare working stock solution of approximately 0.5mg/ml and let the lyophilized pellet dissolve completely. Product is not sterile! Please filter the product by an appropriate sterile filter before using it in
- Zusammensetzung:Filtered (0.4 um) and lyophilized from 0.5 mg/ml in PBS pH 7.4.
- Validierte Anwendungen:SDS-PAGE
Frequently Bought Together