Rat MCP3 (from E. coli)
Anbieter: Biorbyt
Zertifikate
Über diesen Artikel
Technische Daten
- Ursprung:E. coli
- Spezies:Rat
- Lagerbedingungen:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Proteinsynonym:C-C motif chemokine 7
- Protein/Peptid-Name:MCP3
- Purity:> 95% by SDS-PAGE and HPLC analyses
- Molecular weight:8.4 kDa
- Sequence:QPDGTNSSTC CYVKKQKIPK RNLKSYRKIT SSRCPWEAVI FKTKKGMEVCAEAHQKWVEE AIAYLDMKTS TPKP
- Zusammensetzung:Lyophilized from a 0.2 um filtered concentrated solution in 2xPBS, pH 7.4.
- Validierte Anwendungen:SDS-PAGE , HPLC
Technische Daten
Frequently Bought Together