Order Entry
Switzerland
ContactUsLinkComponent
Mouse recombinant IL36RA (from E. coli)
Anbieter:  Biorbyt
Zertifikate
Mouse recombinant IL36RA (from E. coli)
Anbieter:  Biorbyt

Technische Daten

  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Mouse
  • Lagerbedingungen:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Proteinsynonym:
    Murine Interleukin-36 Receptor Antagonist
  • Protein/Peptid-Name:
    IL36RA
  • Purity:
    > 95 % by SDS-PAGE and HPLC analyses
  • Molecular weight:
    16.9 kDa
  • Sequence:
    VLSGALCFRM KDSALKVLYL HNNQLLAGGL HAEKVIKGEE ISVVPNRALD ASLSPVILGVQGGSQCLSCG TEKGPILKLE PVNIMELYLG AKESKSFTFY RRDMGLTSSF ESAAYPGWFLCTSPEADQPV RLTQIPEDPA WDAPITDFYF QQCD
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered concentrated solution in PBS, pH 7.4, 2mM beta -mercaptoethanol
  • Validierte Anwendungen:
    SDS-PAGE , HPLC

Technische Daten

Zugehörige Informationen

Frequently Bought Together