ContactUsLinkComponent
Human recombinant KRT19 (from E. coli)
Human recombinant KRT19 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant KRT19 (from E. coli)
Supplier:  Biorbyt
Specifications
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at −20 °C. to −70 °C. in a manual defrost freezer.Avoid repeated freeze thaw cycles
  • Protein/Peptid-Name:
    KRT19
  • Purity:
    > 95% by SDS-PAGE gel analysis.
  • Zusammensetzung:
    PBS, pH 7.2, sterile filtered
  • Validierte Anwendungen:
    ELISA , WB , DOT , FuncS
Specifications
Related Information
Frequently Bought Together