Order Entry
Switzerland
ContactUsLinkComponent
Human recombinant G-CSF (from bacteria)
Human recombinant G-CSF (from bacteria)
Supplier:  Biorbyt
Certificates
Human recombinant G-CSF (from bacteria)
Supplier:  Biorbyt

Specifications

  • Konjugation:
    Unconjugated
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    Bacteria
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
  • Biologische Aktivität:
    The ED50 was determined by the M1 cell differentiation assay is 0.01 ng/ml, corresponding to specific activity of 100MIU/mg.
  • Gen-ID:
    1440
  • Proteinsynonym:
    Granulocyte ColonyStimulating Factor
  • UniProtKB:
    P09919-2 (Human)
  • Protein/Peptid-Name:
    G-CSF
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MTPLGPASSLPQSFLLKCLEQVRKIQGDGAALQEKLCATYKLCHPEELVLLGHSLGIPWAPLSSC PSQALQLAGCLSQLHSGLFLYQGLLQALEGISPELGPTLDTLQLDVADFATTIWQQMEELGMAPA LQPTQGAMPAFASAFQRRAGGVLVASHLQSFLEVSYRVLRHLAQP
  • Zusammensetzung:
    Lyophilized from a 0.2 um filtered solution of 10mM HAc-NaAc, 150mM NaCl, 0.004% Tween 80, 5% Mannitol, pH 4.0.
  • Validierte Anwendungen:
    SDS-PAGE , FuncS

Specifications

Related Information

Frequently Bought Together