Human recombinant GCA GST (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Konjugation:Unconjugated
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Human
- Lagerbedingungen:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Proteinsynonym:Grancalcin
- UniProtKB:P28676 (Human)
- Protein/Peptid-Name:GCA GST
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MAYPGYGGGFGNFSIQVPGMQMGQPVPETGPAILLDGYSGPAYSDTYSSAGDSVYTYFSAVAGQD GEVDAEELQRCLTQSGINGTYSPFSLETCRIMIAMLDRDHTGKMGFNAFKELWAALNAWKENFMT VDQDGSGTVEHHELRQAIGLMGYRLSPQTLTTIVKRYSKNGRIFFDDYVACCVKLRALTDFFRKR DHLQQGSANFIYDDFLQGTMAI
- Zusammensetzung:Lyophilized from a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 1mM EDTA, pH 8.5.
- Validierte Anwendungen:SDS-PAGE