ContactUsLinkComponent
Human recombinant PDIA6 (from HEK293 cells)
 :  Biorbyt
Human recombinant PDIA6 (from HEK293 cells)
 :  Biorbyt
  • Konjugation:
    Unconjugated
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    HEK293 cells
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Proteinsynonym:
    Protein DisulfideIsomerase A6
  • UniProtKB:
    Q15084 (Human)
  • Protein/Peptid-Name:
    PDIA6
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    LYSSSDDVIELTPSNFNREVIQSDSLWLVEFYAPWCGHCQRLTPEWKKAATALKDVVKVGAVDAD KHHSLGGQYGVQGFPTIKIFGSNKNRPEDYQGGRTGEAIVDAALSALRQLVKDRLGGRSGGYSSG KQGRSDSSSKKDVIELTDDSFDKNVLDSEDVWMVEFYAPWCGHCKNLEPEWAAAASEVKEQTKGK VKLAAVDATVNQVLASRYGIRGFPTIKIFQKGESPVDYDGGRTRSDIVSRALDLFSDNAPPPELL EIINEDIAKRTCEEHQLCVVAVLPHILDTGAAGRNSYLEVLLKLADKYKKKMWGWLWTEAGAQSE LETALGIGGFGYPAMAAINARKMKFALLKGSFSEQGINEFLRELSFGRGSTAPVGGGAFPTIVER EPWDGRDGELPVEDDIDLSDVELDDLGKDELVDHHHHHH
  • Zusammensetzung:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 150mM NaCl, 10% Glycerol, pH 8.0.
  • Validierte Anwendungen:
    SDS-PAGE
Frequently Bought Together