Human recombinant Peroxiredoxin 6 (from E. coli)
Anbieter: Biorbyt
Zertifikate
Über diesen Artikel
Technische Daten
- Konjugation:Unconjugated
- Protein/Peptid-Typ:Rekombinant
- Ursprung:E. coli
- Spezies:Human
- Lagerbedingungen:Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
- Proteinsynonym:Peroxiredoxin6
- UniProtKB:P30041 (Human)
- Protein/Peptid-Name:Peroxiredoxin 6
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:MGSSHHHHHHSSGLVPRGSHMPGGLLLGDVAPNFEANTTVGRIRFHDFLGDSWGILFSHPRDFTP VCTTELGRAAKLAPEFAKRNVKLIALSIDSVEDHLAWSKDINAYNCEEPTEKLPFPIIDDRNREL AILLGMLDPAEKDEKGMPVTARVVFVFGPDKKLKLSILYPATTGRNFDEILRVVISLQLTAEKRV ATPVDWKDGDSVMVLPTIPEEEAKKLFPKGVFTKELPSGKKYLRYTPQP
- Zusammensetzung:Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, pH 7.2.
- Validierte Anwendungen:SDS-PAGE
Technische Daten
Frequently Bought Together