Order Entry
Switzerland
ContactUsLinkComponent
 
Human recombinant Peroxiredoxin 4 (from E. coli)
 :  Biorbyt
 
Human recombinant Peroxiredoxin 4 (from E. coli)
 :  Biorbyt

 

  • Konjugation:
    Unconjugated
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Proteinsynonym:
    Peroxiredoxin4
  • UniProtKB:
    Q13162 (Human)
  • Protein/Peptid-Name:
    peroxiredoxin 4
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MGSSHHHHHHSSGLVPRGSHMWETEERPRTREEECHFYAGGQVYPGEASRVSVADHSLHLSKAKI SKPAPYWEGTAVIDGEFKELKLTDYRGKYLVFFFYPLDFTFVCPTEIIAFGDRLEEFRSINTEVV ACSVDSQFTHLAWINTPRRQGGLGPIRIPLLSDLTHQISKDYGVYLEDSGHTLRGLFIIDDKGIL RQITLNDLPVGRSVDETLRLVQAFQYTDKHGEVCPAGWKPGSETIIPDPAGKLKYFDKLN
  • Zusammensetzung:
    Supplied as a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 1mM DTT, pH 7.2.
  • Validierte Anwendungen:
    SDS-PAGE

 

 

Frequently Bought Together