Human recombinant NgR3 (from HEK293 cells)
Anbieter: Biorbyt
Zertifikate
Über diesen Artikel
Technische Daten
- Konjugation:Unconjugated
- Protein/Peptid-Typ:Rekombinant
- Ursprung:HEK293 cells
- Spezies:Human
- Lagerbedingungen:Store at −20 °C, though stable at room temperature for 3 weeks. Reconstituted protein solution can be stored at 4-7 °C for 2-7 days. Aliquots of reconstituted samples are stable at −20 °C for 3 months.
- Proteinsynonym:Reticulon4 ReceptorLike 1
- UniProtKB:Q86UN2 (Human)
- Protein/Peptid-Name:NgR3
- Purity:>95% as determined by reducing SDS-PAGE.
- Sequence:CPRDCVCYPAPMTVSCQAHNFAAIPEGIPVDSERVFLQNNRIGLLQPGHFSPAMVTLWIYSNNIT YIHPSTFEGFVHLEELDLGDNRQLRTLAPETFQGLVKLHALYLYKCGLSALPAGVFGGLHSLQYL YLQDNHIEYLQDDIFVDLVNLSHLFLHGNKLWSLGPGTFRGLVNLDRLLLHENQLQWVHHKAFHD LRRLTTLFLFNNSLSELQGECLAPLGALEFLRLNGNPWDCGCRARSLWEWLQRFRGSSSAVPCVS PGLRHGQDLKLLRAEDFRNCTGPASPHQIKSHTLTTTDRAARKEHHSPHGPTRSKGHPHGPRPGH RKPGKNCTNPRNRNQISKAGAGKQAPELPDYAPDYQHKFSFDIMPTARPKRKGKCARRTPIRAPS GVQQAVDHHHHHH
- Zusammensetzung:Lyophilized from a 0.2 um filtered solution of 20mM PB, 150mM NaCl, 5% Threhalose, pH 7.2.
- Validierte Anwendungen:SDS-PAGE