ContactUsLinkComponent
Human recombinant BDH2 (from E. coli)
Human recombinant BDH2 (from E. coli)
Supplier:  Biorbyt
Certificates
Human recombinant BDH2 (from E. coli)
Supplier:  Biorbyt
Specifications
  • Konjugation:
    Unconjugated
  • Protein/Peptid-Typ:
    Rekombinant
  • Ursprung:
    E. coli
  • Spezies:
    Human
  • Lagerbedingungen:
    Store at −20 °C, stable for 6 months after receipt. Please minimize freeze-thaw cycles.
  • Proteinsynonym:
    3Hydroxybutyrate Dehydrogenase Type 2
  • UniProtKB:
    Q9BUT1 (Human)
  • Protein/Peptid-Name:
    BDH2
  • Purity:
    >95% as determined by reducing SDS-PAGE.
  • Sequence:
    MGSSHHHHHHSSGLVPRGSHMGRLDGKVIILTAAAQGIGQAAALAFAREGAKVIATDINESKLQE LEKYPGIQTRVLDVTKKKQIDQFASEVERLDVLFNVAGFVHHGTVLDCEEKDWDFSMNLNVRSMY LMIKAFLPKMLAQKSGNIINMSSVASSVKGVVNRCVYSTTKAAVIGLTKSVAADFIQQGIRCNCV CPGTVDTPSLQERIQARGNPEEARNDFLKRQKTGRFATAEEIAMLCVYLASDESAYVTGNPVIID GGWSL
  • Zusammensetzung:
    Supplied as a 0.2 um filtered solution of 20mM TrisHCl, 100mM NaCl, pH 8.0.
  • Validierte Anwendungen:
    SDS-PAGE
Specifications
Related Information
Frequently Bought Together