Rhesus macaque FLT3 ligand (from E. coli)
Supplier: Biorbyt
Certificates
About this item
Specifications
- Ursprung:E. coli
- Spezies:Rhesus monkey
- Lagerbedingungen:Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
- Protein/Peptid-Name:FLT3 Ligand
- Validierte Anwendungen:SDS-PAGE , HPLC