Auftragserfassung
Switzerland
AuftragsverfolgungKontakt
Human MCP1 (from E. coli)

Human MCP1 (from E. coli)

Marketsource
Anbieter:   Biorbyt
Human MCP1 (from E. coli)
Anbieter:  Biorbyt
Marketsource
Zertifikate

2 Varianten

Product Details & Documents

Documents

Spezifikationen für diese Produktfamilie

Technische Daten

Technische Daten

  • Catalog No:
  • BIRBORB80140-5
  • BIRBORB80140-20
  • Protein/Peptid-Name:
  • MCP1
  • MCP1
  • Proteinsynonym:
  • C-C motif chemokine 2
  • C-C motif chemokine 2
  • Spezies:
  • Human
  • Human
  • Ursprung:
  • E. coli
  • E. coli
  • Sequence:
  • The sequence of the first five N-terminal amino acids was determined and was found to be Gln-Pro-Asp-Ala-Ile.
  • The sequence of the first five N-terminal amino acids was determined and was found to be Gln-Pro-Asp-Ala-Ile.
  • Biologische Aktivität:
  • The specific activity as determined by the ability of MCP-1 to chemoattract human Monocytes using concentration of 5-20 ng/ml.
  • The specific activity as determined by the ability of MCP-1 to chemoattract human Monocytes using concentration of 5-20 ng/ml.
  • Purity:
  • > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
  • Zusammensetzung:
  • The protein was lyophilized from concentrated (1mg/ml) sterile solution containing no additives.
  • The protein was lyophilized from concentrated (1mg/ml) sterile solution containing no additives.
  • Lagerbedingungen:
  • Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Löslichkeit:
  • It is recommended to reconstitute the lyophilized Monocyte Chemotactic Protein-1in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • It is recommended to reconstitute the lyophilized Monocyte Chemotactic Protein-1in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
  • Validierte Anwendungen:
  • SDS-PAGE , HPLC , FuncS
  • SDS-PAGE , HPLC , FuncS

Technische Daten

Product Family Options

Produktinformationen

  • VE
    Verfügbarkeit
    Preis
  • 5 µG
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 5 µG
    1 * 5 µG

    Spezifikationen:
    Marketsource
    Mehr
    Specifications
    Cat. No.
    BIRBORB80140-5
    Protein/Peptid-Name
    MCP1
    Proteinsynonym
    C-C motif chemokine 2
    Spezies
    Human
    Ursprung
    E. coli
    Sequence
    The sequence of the first five N-terminal amino acids was determined and was found to be Gln-Pro-Asp-Ala-Ile.
    Biologische Aktivität
    The specific activity as determined by the ability of MCP-1 to chemoattract human Monocytes using concentration of 5-20 ng/ml.
    Purity
    > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
    Zusammensetzung
    The protein was lyophilized from concentrated (1mg/ml) sterile solution containing no additives.
    Lagerbedingungen
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
    Löslichkeit
    It is recommended to reconstitute the lyophilized Monocyte Chemotactic Protein-1in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
    Validierte Anwendungen
    SDS-PAGE , HPLC , FuncS
    Marketsource
  • 20 µG
    Einloggen für Verfügbarkeit
    Einloggen für Verfügbarkeit
    1 * 20 µG
    1 * 20 µG

    Spezifikationen:
    Marketsource
    Mehr
    Specifications
    Cat. No.
    BIRBORB80140-20
    Protein/Peptid-Name
    MCP1
    Proteinsynonym
    C-C motif chemokine 2
    Spezies
    Human
    Ursprung
    E. coli
    Sequence
    The sequence of the first five N-terminal amino acids was determined and was found to be Gln-Pro-Asp-Ala-Ile.
    Biologische Aktivität
    The specific activity as determined by the ability of MCP-1 to chemoattract human Monocytes using concentration of 5-20 ng/ml.
    Purity
    > 98.0% as determined by RP-HPLC and analysis by SDS-PAGE
    Zusammensetzung
    The protein was lyophilized from concentrated (1mg/ml) sterile solution containing no additives.
    Lagerbedingungen
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
    Löslichkeit
    It is recommended to reconstitute the lyophilized Monocyte Chemotactic Protein-1in sterile H2O not less than 100ug/ml, which can then be further diluted to other aqueous solutions.
    Validierte Anwendungen
    SDS-PAGE , HPLC , FuncS
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.