ContactUsLinkComponent
rHBsAgadr (from CHO cells)
Anbieter:  Biorbyt
Zertifikate
rHBsAgadr (from CHO cells)
Anbieter:  Biorbyt
Technische Daten
  • Ursprung:
    CHO cells
  • Lagerbedingungen:
    Store at 4 °C for up to two weeks. For long term storage, aliquot and store at −20 °C, avoid freeze/thaw cycles.
  • Proteinsynonym:
    Hepatitis B Surface Antigen
  • Protein/Peptid-Name:
    rHBsAgadr
  • Purity:
    > 95% by HPLC analyses.
  • Molecular weight:
    23.0-27.0 kDa
  • Zusammensetzung:
    Sterile Filtered solution containing 20 mM PB, 154 mM sodium chloride.
  • Validierte Anwendungen:
    SDS-PAGE , HPLC
Technische Daten
Zugehörige Informationen
Frequently Bought Together