GC Columns, Rtx-Volatile Amine (fused silica)
Anbieter: Restek
Zertifikate
Über diesen Artikel
The Rtx-Volatile amine column was designed for analyzing volatile amines in difficult matrices, such as water.
- Unique selectivity for baseline resolution of all volatile amines
- Excellent inertness assures accuracy and sensitivity for volatile amines, including free ammonia
- Highly robust phase withstands repeated water injections, resulting in longer column lifetime
- High temperature stability (290 °C) ensures elution of amines up to C16 and allows contaminants to be removed by 'baking out' the column
Frequently Bought Together