Detergents for glassware washers, ProCare Lab 11 MA
MIEL10266720
: MIELE
- Kapazität:10 kg
- Bezeichnung:Alkaline detergent powder, ProCare Lab 11 MA
- Environmentally Preferable:
- VE:10 kg
- pH-Bereich:11,7 - 11,9
Powder, mildly alkaline, foam-retardant action detergent for machine based cleaning of laboratory glassware and laboratory utensils.
- Provides good protection to surfaces
- Removes blood, protein and food residues
- Free from oxidative agents
ProCare Lab 11 MA is for general-purpose use in medical, biological and chemical laboratories.
Note: Suitable for stainless steel, laboratory glassware, ceramics, plastics such as PE, PP, PVDF, PTFE. Conditionally suitable for use on non ferrous metals and aluminium. Not suitable for anodised aluminium.