Survival Motor Neuron (SMN) ELISA Kits
Katalognr. ENZOADI900209
Anbieter: ENZO LIFE SCIENCES
CAS-Nummer:
Technische Daten
- VE:1 KIT
- Bezeichnung:SMN ELISA kit, 96 wells
- Seq:02
Technische Daten
Über diesen Artikel
The SMN ELISA Kit is a colorimetric immunometric immunoassay kit for the quantitative determination of human and mouse SMN in cell lysate samples.
- Highly sensitive measurement, detecting as little as 50 pg/ml of SMN
- High throughput format with results in <3 hours for up to 39 samples in duplicate
- Fully quantitative results that surpass semi-quantitative Western blot analysis
- Convenient - ready to use liquid colour coded reagents
- Pre-coated plate saves time and minimise errors
Delivery: The kit contains microtiter plate, conjugate, antibody, assay buffer 13, wash buffer concentrate, standard, TMB substrate, stop solution 2, and extraction reagent 4.