Anti-BMPR1A Mouse Monoclonal Antibody [clone: 18B7]
Codice catalogo GNTXGTX11490
Nome del fornitore: Genetex
Specifiche
- Antikörper-Typ:Primär
- Antigen-Name:Bone morphogenetic Protein Receptor, type IA
- Antigen-Symbol:BMPR1A
- Klonalität:Monoklonal
- Klon:18B7
- Konjugation:Unconjugated
- ELISA:Yes
- Wirt:Mouse
- Isotyp:IgG1
- Reaktivität:Human
- Western blot:Yes
- Gen-ID:796
- Antigen-Synonyme:serine/threonine-protein kinase receptor R5|activin A receptor|BMPR-1A|SKR5|BMP type-1A receptor|ALK-3|bone morphogenetic protein receptor type-1A|ALK3|type II-like kinase 3|10q23del|CD292|ACVRLK3|activin receptor-like kinase 3|BMPRIA
- Lagerungspuffer:Phoshate-buffered saline, pH 7,4, containing 0,1% sodium azide
- Lagertemperatur:Keep as concentrated solution, aliquot and store at 4 °C.
- Immunogen:Full length protein (Human).
- Reinigung:Protein A purified
- Größe:100 μg
- VE:100 µG
Specifiche
Informazioni su questo articolo
Type: Primary
Antigen: BMPR1A
Clonality: Monoclonal
Clone: Unconjugated
Conjugation: 18B7
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human