Rat CRP/C reactive protein ELISA kit
Catalog # BIRBORB79257-96
Supplier: Biorbyt
CAS Number:
Specifications
- VE:96
- Reaktivität der primären Antikörper:Rat
- Bezeichnung:Rat C Reactive Protein ELISA Kit
Specifications
About this item
Rat C Reactive Protein (CRP) ELISA Kit is used for the quantitative determination of rat C reactive protein in serum, plasma or cell culture supernatant.