Anti-ABHD5 Rabbit Polyclonal Antibody
100 μg
Catalog # BSBTPB10021
Supplier: Boster Bio
Jump to:
Product Details & Documents
Rabbit IgG polyclonal antibody for 1-acylglycerol-3-phosphate O-acyltransferase ABHD5(ABHD5) detection. Tested with WB, IHC-P in Human;Rat.
Type: Primary
Antigen: ABHD5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Documents
PB10021
Datasheet
Specifications
- Größe:100 μg
- Western blot:Yes
- Antikörper-Typ:Primär
- Konjugation:Unconjugated
- Reaktivität:Human|Rat
- Kreuzadsorption:No
- Cat. No.:BSBTPB10021
- Antigen-Name:Abhydrolase Domain Containing 5
- Gen-ID:51099
- Antigen-Synonyme:NCIE2|IECN2|CDS|CGI58
- Antigen-Symbol:ABHD5
- Klonalität:Polyklonal
- Isotyp:IgG
- Wirt:Rabbit



