Digitales Rotationsviskosimeter Modell DV-II+ Pro
Supplier: AMETEK
Certificates
About this item
The Extra DV2+ PRO features a ball bearing suspension system.
- EZ-Lock spindle coupling, another productivity booster that allows to attach/detach spindles
- Quick Action Lab Stand allows users to move the viscometer up and down the stand
- Cones/plate versions are also available
Specially designed for busy environments.
Ordering information: Includes a case, a stand and a spindle set, but also a temperature probe, DV Loader software and calibration certificate.
Frequently Bought Together