ContactUsLinkComponent
(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure 97%
  ACRO434490010
 :  
undefined
(R)-(-)-2-Fluor-α-methyl-4-biphenylessigsäure 97%
  ACRO434490010
 :  
  • Packung:
    Glasflasche
  • VE:
    1 g
  • Seq:
    3047
  • Appearance:
    White to off-white Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    106 to 110 °C
  • HPLC:
    ≥96.0 %
  • Specific optical rotation:
    -35° to -45° (20 °C, 589 nm) (c=1, chloroform)
 :51543-40-9
 :
 :C₁₅H₁₃FO₂
 :MFCD00869714
 :244.260 g/mol
 :106…110 °C
 :Raumtemperatur
 :2811
 :6.1,II
 :GHS06