Specifications
- Antikörper-Typ:Kontrolle
- Antigen-Symbol:IgG2a kappa
- Klon:RTK2758
- Konjugation:PE (Phycoerythrin)/Dazzle 594
- Wirt:Rat
- Immunogen:Trinitrophenol + KLH
- VE:100 µG
Specifications
About this item
The RTK2758 immunoglobulin is useful as an isotype-matched control (for the relevant formats) for Western blotting, immunoprecipitation, immunohistochemistry, functional assay, immunofluorescence microscopy, immunocytochemistry and immunofluorescent staining (surface or intracellular) for flow cytometric analysis. The LEAF™ purified antibody (Endotoxin <0.1 EU/μg, Azide-Free, 0.2 μm filtered) is recommended for functional assays as negative control. For in vivo studies or highly sensitive assays, we recommend Ultra-LEAF™ purified antibody with a lower endotoxin limit than standard LEAF™ purified antibodies (Endotoxin <0.01 EU/µg).
Type: Control
Antigen: IgG2a kappa
Clonality:
Clone: RTK2758
Conjugation: PE/Dazzle 594
Epitope:
Host: Rat
Isotype:
Reactivity: