ContactUsLinkComponent
Protein transfection reagent, Pierce™
Katalognr. PIER89850
CAS-Nummer:  
undefined
Protein transfection reagent, Pierce™
Katalognr. PIER89850
CAS-Nummer:  
Technische Daten
  • VE:
    1 KIT
  • Protein delivery:
    Ja
  • Bezeichnung:
    Protein transfection reagent, 250 µl
Technische Daten
Über diesen Artikel
This Pierce™ protein transfection reagent is a cationic lipid mixture for complexation with proteins, peptides, antibodies and other biologically active molecules to allow their direct intracellular delivery.
  • No need to isolate, clone and transfect gene sequences thu saving time
  • Optimum delivery in three to four hours after incubation
  • Delivery complex is non-covalent with proteins and peptides retaining biological activity after delivery
  • Works with proteins, peptides and fluorescently labeled antibodies making it versatile
Molecules that have been successfully delivered with the transfection reagent include β-galactosidase, fluorescent-antibody, green fluorescent protein (GFP), high and low molecular weight dextran sulfate, phycoerythrin-BSA, caspase 3, caspase 8 and granzyme B.