Bulb for pipettes, SP Bel-Art
BELAH378880000
- Bezeichnung:Safety bulb for pipettes
- VE:1
Pipette bulb made of silicone rubber.
- Tapered, silicone rubber seal provides an airtight fit on all size pipettes
- Can be opened, cleaned and steam autoclaved at 121 °C
- Provided with an elastic cord for dedicating that pipettor to a specific reagent bottle
- For use with Westergren or Wintrobe sedimentation tubes