DAPT
10 mg
Anbieter: Thermo Fisher Scientific
Direkt zu
Product Details & Documents
CAS No.: 208255-80-5 Molecular Formula: C23H26F2N2O4 Formula Weight: 432.50 Storage Temperature: -30°C to -10°C Physical Form: Lyophilized Sensitivity: Light Sensitive Appearance: White MDL No.: MFCD04974585
Applications: A cell-permeable dipeptide that suppresses A production by blocking -secretase
Synonyme: N-[N-(3,5-Difluorophenacetyl-L-alanyl)]-S-phenylglycine t-Butyl Ester, tert-Butyl (S)-2-((S)-2-(2-(3,5-difluorophenyl)acetamido)propanamido)-2-phenylacetate, (3,5-Difluorophenylacetyl)-Ala-Phg-OtBu, (3,5-Difluorophenylacetyl)-L-alanyl-L-2-phenylglycine tert-Butyl Ester
- CAS Nummer:208255-80-5
- Formel:C₂₃H₂₆F₂N₂O₄
- MDL-Nummer:MFCD04974585
- Molekulargewicht:432.47 g/mol
Documents
Technische Daten
Technische Daten
- VE:10 mg
Technische Daten
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



