Ebastin
Catalog # J63088.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- VE:1 g
- Seq:3047
Specifications
About this item
CAS Number:90729-43-4
:
Formula:C₃₂H₃₉NO₂
MDL Number:MFCD00865661
:469.67 g/mol
Merck Index:14,3484
Melting Pt:84…87 °C
Storage Temperature:Raumtemperatur