Zinkoxid 50% in 1,2-Propandiolmonomethyletheracetat, kolloidale Dispersion mit Detergens, NanoArc® ZN-0625
Catalog # 44924.22
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- VE:100 g
- Seq:3401
Specifications
About this item
:
:81.39 g/mol
Storage Temperature:Raumtemperatur
UN:1993
ADR:3,III
:GHS02
CAS Number:1314-13-2
Formula:ZnO
MDL Number:MFCD00011300