Anti-FIGF Rabbit Polyclonal Antibody
Catalog # BSBTPB9869
Supplier: Boster Bio
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:c-fos induced growth factor (vascular endothelial growth factor D)
- Antigen-Symbol:FIGF
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human
- Western blot:Yes
- Kreuzadsorption:No
- Gen-ID:2277
- Antigen-Synonyme:VEGF-D|VEGFD
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Type: Primary
Antigen: FIGF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human