Mouse IgG1 kappa Isotype Control [clone: MOPC-21] (Alexa Fluor® 594)
Catalog # BLEG400174
Supplier: Biolegend
Specifications
- Antikörper-Typ:Kontrolle
- Antigen-Symbol:IgG1 kappa
- Klonalität:Monoklonal
- Klon:MOPC-21
- Konjugation:Alexa Fluor® 594
- Wirt:Mouse
- ImmunFluoreszenz:Yes
- Isotyp:IgG1 kappa
- Reaktivität:Mouse
- Form:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide.
- Lagertemperatur:The antibody solution should be stored undiluted between 2°C and 8 °C, and protected from prolonged exposure to light. Do not freeze.
- Konzentration:0,5 mg/ml
- Transporttemperatur:Ambient RT
- Reinigung:The antibody was purified by affinity chromatography and conjugated with Alexa Fluor® 594 under optimal conditions. The solution is free of unconjugated Alexa Fluor® 594.
- Größe:100 µg
- VE:100 µG
Specifications
About this item
Type: Control
Antigen: IgG1 kappa
Clonality: Monoclonal
Clone: MOPC-21
Conjugation: Alexa Fluor® 594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse