ContactUsLinkComponent
Reagent reservoirs
Katalognr. 613-3093
Reagent reservoirs
Katalognr. 613-3093
Technische Daten
  • Verpackt:
    Individually wrapped
  • Steril:
    +
  • Typ:
    V-bottom
  • Kapazität:
    25 ml
  • Farbe:
    Weiß
  • Seq:
    01
  • VE:
    100
Technische Daten
Über diesen Artikel
These reagent reservoirs made of PS or PVC are designed to accommodate all types of pipettors from single channel to 16-channel.
  • Made of FDA grade plastic
  • Graduation marks on interior wall
  • Lot certified Rnase/Dnase-free and non pyrogenic
  • Pour-off spouts in each corner
  • Availabe non sterile or sterile; sterility validated at 10⁻⁶