Brewers QCheck® kit
Katalognr. 710-1968
Anbieter: DOEHLER GROUP
CAS-Nummer:
Technische Daten
- Packung:Bottle
- VE:1
- Bezeichnung:Brewers QCheck® Kit
Technische Daten
Über diesen Artikel
The Brewers QCheck® Kit contains ready-to-use culture media, allowing simple, fast and comprehensive microbiological control of all samples in the brewing process. The kit is particularly suitable for micro- and craft breweries who wish to establish microbiological quality control in their processes. All microbiological findings can be easily and reliably seen through a colour change, even without any previous knowledge. Only minimal laboratory equipment is required. The Brewers QCheck® Kit thus allows the entire beer brewing process to be microbiologically controlled, providing the perfect introduction to microbiological quality control.
- Ready to use - easy-to-use culture media without production
- Everything you need - complete kit including illustrated manual
- Simple evaluation - clear and easily visible colour change
- Reliable findings - fully tested, optimised medium composition
Delivery: The kit contains: LMC for water analysis, 2×250 ml in a carrier bag; NBB®-B tubes for yeast and beer analysis 20×10 ml in a box; NBB®-B-Am tubes for hygiene monitoring in production, filling and dispensing units 20×10 ml in a box. Sterile swabs for sampling 3×10 sterile swabs (individually packaged).