Anti-Bag3 Rabbit Polyclonal Antibody
BSBTPB9754
- Antikörper-Typ:Primär
- Antigen-Symbol:BAG3
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- ImmunoChemie:Yes
- Isotyp:IgG
- Reaktivität:Human,Rat
- Western blot:Yes
- Kreuzadsorption:No
- Gen-ID:9531
- Größe:100 µg
- VE:100 µG
Rabbit IgG polyclonal antibody for BAG family molecular chaperone regulator 3(BAG3) detection. Tested with WB, IHC-P in Human;Rat.
Type: Primary
Antigen: Bag3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat