Functional ear model
Functional ear model
Katalognr. BINH1005052
Marketsource
Anbieter: 3B Scientific
Direkt zu
Product Details & Documents
This functional ear model shows how the tympanic membrane, ossicles, the complex internal ear with the cochlea and the oscillations of the basilar hearing membrane operate/interact.
- Enclosed mirror enables operation of the functional ear model
- One single ear model may be studied by several students simultaneously in an action-oriented learning situation
Suitable for the studying of various ear-functions from different angles at the same time.
Ordering information: Functional ear includes a four-colour explanatory chart.
Technische Daten
- Bezeichnung:Functional ear model