Venous access device model
Torso Venous Access Model
Katalognr. BINH1005563
Marketsource
Anbieter: 3B Scientific
Direkt zu
Product Details & Documents
Teach patients and healthcare staff the care and use of two different types of catheters with this model. Features left and right subclavian catheter connections that may be flushed with water; implanted venous access system with a 'skin' flap over the implant to allow viewing.
- Mounted on a collapsible easel
- Made of skinned and lightweight urethane foam
- May be washed with soap, water as well as antiseptics
Verpackung: In its carrying case.
Technische Daten
- Bezeichnung:Torso Venous Access Model