Accessories for chromatography systems
: Cytiva
Product Family Options
- BezeichnungVE
- Union fingertight, PEEK, Int.Ø 0,3 mm41 * 4 STSFr. 156.00
- Tubing connector luer lock, 5/16" female, PP41 * 4 STSFr. 96.00
- Tubing connector flangeless, M6 female, PP21 * 2 STSFr. 85.00
- Tubing connector flangeless, M6 female, PP21 * 2 STSFr. 92.20
- O-ring for inner end piece to superloop, EPDM, 150 ml21 * 2 STSFr. 44.50
- Tube for super loop, 150 ml, borosilicate glass11 * 1 STSFr. 542.00
- Column filter, for use with fixed volume column reactor 24 ml21 * 2 STSFr. 235.00
- Tubing, FEP, 3 m, Int.Ø 2,9 mm, Ext.Ø 4,76 mm11 * 1 STSFr. 123.00
- pH flow cell, including dummy electrode, round tip, flow rate 100 ml/min11 * 1 STSFr. 1’370.00
- Inlet tubing kit, FEP, Int.Ø 2,9 mm, Ext.Ø 4,76 mm11 * 1 STSFr. 205.00
- Inlet filter set, 10 filters/nets, PP, Ext.Ø 25 mm10 SET1 * 10 SETSFr. 223.00
- Union, M6 female for tubing ¹/₁₆", PEEK21 * 2 STSFr. 171.00
- New purge kit, includes tubing of length 160 mm with luer union1 KIT1 * 1 KITSFr. 98.00
- Tubing, PEEK, 2 m, Int.Ø 0,15 mm, ext.Ø 1,58 mm, purple1 KIT1 * 1 KITSFr. 603.00
- O-ring, 64,5×3 mm, EPDM glass tube 6021 * 2 STSFr. 522.00
- Adjusting inner rod, acetal plastic (POM)11 * 1 STSFr. 119.00
- Unflanged tubing, 4 m, PTFE, Int.Ø 1,9 mm, Ext.Ø 2,7 mm71 * 7 STSFr. 589.00
- S-flanged tubing, PTFE, flanged in one end with M6 male connector, 2 m, Int.Ø 1,9 mm, Ext.Ø 2,7 mm11 * 1 STSFr. 134.00
- Pre-flanged tubing, 1 m, Int.Ø 1,9 mm, Ext.Ø 2,7 mm21 * 2 STSFr. 177.00
This product has been blocked by your organization. Please contact your purchasing department for more information