Anti-Peroxiredoxin 5 Rabbit Polyclonal Antibody
Katalognr. ABFRLF-PA0010
Anbieter: AbFrontier
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:Peroxiredoxin 5
- Antigen-Symbol:PRDX5
- Klonalität:Polyklonal
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Mouse
- Western blot:Yes
- Lagertemperatur:Store for 1 year at –20 °C from date of shipment
- Konzentration:1 mg/ml
- Immunogen:Recombinant human fragment protein(without mitochondrial leader sequence) purified from E.coli
- Reinigung:Protein A column
- Größe:100 µl
- VE:100 µl
Technische Daten
Über diesen Artikel
Type: Primary
Antigen: Peroxiredoxin 5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse