Anti-TMBIM6 Mouse Monoclonal Antibody [clone: 20F248]
Catalog # ABNOMAB0106
Supplier: Abnova
Specifications
- Antikörper-Typ:Primär
- Antigen-Name:Transmembrane BAX Inhibitor Motif Containing 6
- Antigen-Symbol:TMBIM6
- Klonalität:Monoklonal
- Klon:20F248
- Konjugation:Unconjugated
- Wirt:Mouse
- Isotyp:IgM
- Reaktivität:Human
- Western blot:Yes
- Kreuzadsorption:No
- Format:Liquid
- Gen-ID:7009
- Antigen-Synonyme:BI-1|TEGT|BI1|Bax inhibitor 1|Testis-enhanced gene transcript protein,Transmembrane BAX inhibitor motif-containing protein 6
- Lagerungspuffer:PBS (0,05% sodium azide)
- Lagertemperatur:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A mixture of synthetic peptides corresponding to N-terminus, internal and C-terminuss of human TMBIM6.
- Größe:200 μl
- VE:200 µl
Specifications
About this item
Mouse monoclonal antibody raised against synthetic peptide of TMBIM6.
Type: Primary
Antigen: TMBIM6
Clonality: Monoclonal
Clone: 20F248
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human