Anti-IGFR3 Mouse Monoclonal Antibody (Unconjugated) [clone: ]
Type: Primary
Antigen: CD16
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Baboon, Chimpanzee, Cynomolgus, Human, Marmoset, Rhesus
- Catalog No:
- BLEG302001
- BLEG302002
- Antigen-Symbol:
- IGFR3
- IGFR3
- Antigen-Name:
- IgG Fc Receptor 3
- IgG Fc Receptor 3
- Antigen-Synonyme:
- CD16|FCRIII|CD16A|FCGR3|IMD20|FCG3|FCRIIIA|FCR-10|FCGRIII|IGFR3
- CD16|FCRIII|CD16A|FCGR3|IMD20|FCG3|FCRIIIA|FCR-10|FCGRIII|IGFR3
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Monoklonal
- Monoklonal
- Konjugation:
- Unconjugated
- Unconjugated
- Reaktivität:
- Chimpanzee, Human
- Human
- Wirt:
- Mouse
- Mouse
- Isotyp:
- IgG1 kappa
- IgG1 kappa
- ImmunoChemie:
- Yes
- Yes
- Immunpräzipitation:
- Yes
- Yes
- Durchflusszytometrie:
- Yes
- Yes
Frequently Bought Together