Anti-LY6G Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: ]
Anbieter: Biolegend
Zertifikate
Über diesen Artikel
Type: Primary
Antigen: LY6G
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Technische Daten
- Catalog No:
- BLEG127606
- BLEG127605
- Antigen-Symbol:
- LY6G
- LY6G
- Antigen-Name:
- Lymphocyte Antigen 6 Complex Locus G
- Lymphocyte Antigen 6 Complex Locus G
- Antigen-Synonyme:
- Gr1|Gr-1|Ly-6G
- Gr1|Gr-1|Ly-6G
- Antikörper-Typ:
- Primär
- Primär
- Klonalität:
- Monoklonal
- Monoklonal
- Konjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reaktivität:
- Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Baboon, Cynomolgus monkey, Rhesus monkey, Chimpanzee, Human
- Wirt:
- Rat
- Rat
- Isotyp:
- IgG2a kappa
- IgG2a kappa
- Durchflusszytometrie:
- Yes
- Yes
Technische Daten
Frequently Bought Together