Order Entry
Switzerland
ContactUsLinkComponent
Anti-ORAI1 Mouse Monoclonal Antibody [clone: 6D11A11]
Anti-ORAI1 Mouse Monoclonal Antibody [clone: 6D11A11]
Catalog # ORIGTA320060
Supplier:  OriGene
Anti-ORAI1 Mouse Monoclonal Antibody [clone: 6D11A11]
Catalog # ORIGTA320060
Supplier:  OriGene

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Symbol:
    ORAI1
  • Klonalität:
    Monoklonal
  • Klon:
    6D11A11
  • Konjugation:
    Unconjugated
  • Wirt:
    Mouse
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG2b
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Kreuzadsorption:
    No
  • Form:
    Liquid
  • Lagerungspuffer:
    ORAI1 Monoclonal Antibody is supplied in PBS containing 0,02% sodium azide.
  • Lagertemperatur:
    –20 °C
  • Transporttemperatur:
    –20 °C
  • Immunogen:
    Mouse monoclonal ORAI1 antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human ORAI1.
  • Reinigung:
    purified by immunoaffinity chromatography
  • Größe:
    100 μg
  • VE:
    100 µG

Specifications

About this item

Recommended Dilutions: Western Blot: 1-2 ?g/ml; Immunochemistry: 2.5 ?g/ml; Immunofluorescence: 20 ?g/ml

Type: Primary
Antigen: ORAI1
Clonality: Monoclonal
Clone: 6D11A11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human, Mouse, Rat