Anti-SLC2A2 Rabbit Polyclonal Antibody
Product Details & Documents
Rabbit IgG polyclonal antibody for Solute carrier family 2, facilitated glucose transporter member 2(SLC2A2) detection. Tested with WB, IHC-P in Human.
SLC2A2, also known as Glucose transporter 2 (GLUT2), is a transmembrane carrier protein that enables protein facilitatedglucose movement across cell membranes. This gene encodes an integral plasma membrane glycoprotein of the liver, islet beta cells, intestine, and kidney epithelium. The encoded protein mediates facilitated bidirectional glucose transport. Because of its low affinity for glucose, it has been suggested as a glucose sensor. Mutations in this gene are associated with susceptibility to diseases, including Fanconi-Bickel syndrome and noninsulin-dependent diabetes mellitus (NIDDM). Alternative splicing results in multiple transcript variants of this gene.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for SLC2A2 is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
By Heat: Boiling the paraffin sections in 10mM citrate buffer, pH6.0, for 20mins is required for the staining of formalin/paraffin sections.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: SLC4A1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Dokumente
PB9436
Technisches Datenblatt
Technische Daten
- Größe:100 µg/vial
- Western blot:Yes
- Antikörper-Typ:Primär
- Form:Lyophilized
- Reinigung:Immunogen affinity purified.
- UniProtKB:P11168
- Reaktivität:Human
- ImmunoChemie:Yes
- Cat. No.:BSBTPB9436
- Antigen-Name:Solute carrier family 2 (facilitated glucose transporter), member 2
- Lagertemperatur:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Gen-ID:6514
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human SLC2A2 (486-514aa ETKGKSFEEIAAEFQKKSGSAHRPKAAVE), different from the related mouse sequence by five amino acids, and from the related rat sequence by seven amino acids.
- Antigen-Synonyme:GLUT2
- Antigen-Symbol:SLC2A2
- Klonalität:Polyklonal
- Isotyp:IgG
- Wirt:Rabbit



