Order Entry
Switzerland
ContactUsLinkComponent
Anti-Sonic Hedgehog Rabbit Antibody
  BSBTPA1072-1
Anti-Sonic Hedgehog Rabbit Antibody
  BSBTPA1072-1

 

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Sonic Hedgehog
  • Antigen-Symbol:
    SHH
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    N-Terminal
  • Kreuzadsorption:
    No
  • Form:
    Lyophilized
  • Antigen-Synonyme:
    TPT|HLP3|HHG1|SMMCI|HPE3|MCOPCB5|TPTPS
  • Aminosäurezahl:
    203 to 218
  • Sequence:
    ATVHLEQGGTKLVKDL
  • Lagertemperatur:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the N-terminus of human Sonic Hedgehog (203-218aa ATVHLEQGGTKLVKDL), identical to the related rat and mouse sequences.
  • Reinigung:
    Purified by immunoaffinity chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

 

 

Rabbit IgG polyclonal antibody for Sonic hedgehog protein(SHH) detection. Tested with WB, IHC-P in Human;Mouse;Rat.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: Sonic Hedgehog
Clonality:
Clone:
Conjugation:
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat