Order Entry
Switzerland
ContactUsLinkComponent
Anti-IRF1 Rabbit Antibody
Katalognr. BSBTPA2237
Anbieter:  Boster Bio
undefined
Anti-IRF1 Rabbit Antibody
Katalognr. BSBTPA2237
Anbieter:  Boster Bio

Technische Daten

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    IRF1
  • Antigen-Symbol:
    IRF1
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • ImmunoChemie:
    Yes
  • Isotyp:
    IgG
  • Reaktivität:
    Human
  • Western blot:
    Yes
  • Kreuzadsorption:
    No
  • Form:
    Lyophilised
  • Aminosäurezahl:
    169 to 183
  • Sequence:
    YMQDLEVEQALTPAL
  • Lagertemperatur:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence in the middle region of human IRF1 (169-183aa YMQDLEVEQALTPAL).
  • Reinigung:
    Purified by immunoaffinity chromatography
  • Größe:
    100 µg
  • VE:
    100 µG

Technische Daten

Über diesen Artikel

Rabbit IgG polyclonal antibody for Interferon regulatory factor 1(IRF1) detection. Tested with WB, IHC-P, ICC in Human.

Reconstitute with distilled water.

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: IRF1
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human