Order Entry
Switzerland
ContactUsLinkComponent
Anti-ITGA7 Rabbit Antibody
Anti-ITGA7 Rabbit Antibody
Catalog # BSBTPA2226
Supplier:  Boster Bio
undefined
Anti-ITGA7 Rabbit Antibody
Catalog # BSBTPA2226
Supplier:  Boster Bio

Specifications

  • Antikörper-Typ:
    Primär
  • Antigen-Name:
    Integrin Alpha 7
  • Antigen-Symbol:
    ITGA7
  • Klonalität:
    Polyklonal
  • Konjugation:
    Unconjugated
  • Wirt:
    Rabbit
  • Isotyp:
    IgG
  • Reaktivität:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Epitop:
    near the C-terminus
  • Kreuzadsorption:
    No
  • Form:
    Lyophilised
  • Antigen-Synonyme:
    Integrin alpha-7 light chain|Integrin alpha-7|UNQ406/PRO768|Integrin alpha-7 70 kDa form|ITGA7
  • Aminosäurezahl:
    1123 - 1139
  • Sequence:
    HAVKIPREDRQQFKEEK
  • Lagertemperatur:
    –20 °C
  • Immunogen:
    A synthetic peptide corresponding to a sequence at the C-terminus of human ITGA7 (1123-1139aa HAVKIPREDRQQFKEEK), identical to the related mouse sequence, and different from the related rat sequence by one amino acid.
  • Reinigung:
    purified by immunogen affinity purification
  • Größe:
    100 µg
  • VE:
    100 µG

Specifications

About this item

Rabbit IgG polyclonal antibody for Integrin alpha-7(ITGA7) detection. Tested with WB in Human, Mouse, Rat.

Reconstitute with distilled water

Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.

Type: Primary
Antigen: ITGA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat