Anti-NFkB p105/p50 Rabbit Antibody
Katalognr. BSBTPA2111-1
Anbieter: Boster Bio
Technische Daten
- Antikörper-Typ:Primär
- Antigen-Name:Nuclear factor NF-kappa-B p105 subunit
- Antigen-Symbol:NFkB p105/p50
- Klonalität:Polyklonal
- Konjugation:Unconjugated
- Wirt:Rabbit
- Isotyp:IgG
- Reaktivität:Human,Rat,Mouse
- Western blot:Yes
- Epitop:near the N-terminus
- Kreuzadsorption:No
- Form:Lyophilised
- Antigen-Synonyme:EBP-1|Nuclear factor NF-kappa-B p50 subunit|Nuclear factor NF-kappa-B p105 subunit|NFKB1|DNA-binding factor KBF1|Nuclear factor of kappa light polypeptide gene enhancer in B-cells 1
- Aminosäurezahl:193 - 208
- Sequence:KELIRQAALQQTKEMD
- Lagertemperatur:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the N-terminus of human NFkB p105 (193-208aa KELIRQAALQQTKEMD), different from the related rat sequence by one amino acid and from the related mouse sequence by two amino acids.
- Reinigung:purified by immunogen affinity purification
- Größe:100 µg
- VE:100 µG
Technische Daten
Über diesen Artikel
Rabbit IgG polyclonal antibody for Nuclear factor NF-kappa-B p105 subunit(NFKB1) detection. Tested with WB in Human, Mouse, Rat.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: NFkB p105/p50
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: near the N-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat